Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Antibody (C19/366), Novus Biologicals™

Mouse Monoclonal Antibody
$536.00
Specifications
Antigen | CD19 |
---|---|
Clone | C19/366 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 0.5-1.0ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CD19 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
P15391 | |
930 | |
Recombinant full-length human CD19 protein (Uniprot: P15391) | |
Primary | |
Store at 4C. |
C19/366 | |
Immunohistochemistry-Paraffin 0.5-1.0ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
CD19 | |
IgG1 κ | |
Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title