Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19 Antibody (CB19) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 9 publications

$207.00 - $524.00

Specifications

Antigen CD19
Clone CB19
Concentration 1.0 mg/mL
Dilution Western Blot reported in scientific literature (PMID 17327405), Flow Cytometry 0.2 ug/10^6 cells, Immunocytochemistry/Immunofluorescence 1:25-1:100, In vitro assay reported in scientific literature (PMID 9576485), CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, In vitro Assay
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP225196SS
SDP
View Documents
Novus Biologicals
NBP225196SS
0.025 mg
Each for $207.00
Only null left
Add to Cart
 
NBP225196
SDP
View Documents
Novus Biologicals
NBP225196
0.1 mg
Each for $524.00
Only null left
Add to Cart
 
Description

Description

CD19 Monoclonal specifically detects CD19 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, In vitro assay, CyTOF-ready.
Specifications

Specifications

CD19
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, In vitro Assay
Unconjugated
Mouse
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers
P15391
930
Intact normal human B cell cells were used as immunogen to generate the CB19 clone.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
CB19
Western Blot reported in scientific literature (PMID 17327405), Flow Cytometry 0.2 ug/10^6 cells, Immunocytochemistry/Immunofluorescence 1:25-1:100, In vitro assay reported in scientific literature (PMID 9576485), CyTOF-ready
Monoclonal
Purified
RUO
Human, Mouse
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
CD19
IgG1
Protein G purified
This product reacts with Human.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.