Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Antibody (CVID3/155), Novus Biologicals™

Mouse Monoclonal Antibody
$280.00 - $536.00
Specifications
Antigen | CD19 |
---|---|
Clone | CVID3/155 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, SDS-Page, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24467400
![]() |
Novus Biologicals
NBP2446740.02MG |
0.02 mg |
Each for $280.00
|
|
|||||
NBP24467401
![]() |
Novus Biologicals
NBP2446740.1MG |
0.1 mg |
Each for $536.00
|
|
|||||
NBP24467402
![]() |
Novus Biologicals
NBP2446740.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD19 Monoclonal specifically detects CD19 in Human, Monkey samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CD19 | |
0.2 mg/mL | |
Flow Cytometry, SDS-Page, Immunofluorescence | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
930 | |
Recombinant human CD19 protein | |
Primary | |
Store at 4C. | |
95 kDa |
CVID3/155 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Monkey | |
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
CD19 | |
IgG1 κ | |
Protein A or G purified | |
Recognizes a protein of 95kDa, identified as CD19. CD19 is a transmembrane glycoprotein that contains two extracellular immunoglobulin-like domains. CD19 is present in both benign and malignant B-cells and is considered to be the most reliable surface marker of this lineage over a wide range of maturational stages. In normal lymphoid tissue, CD19 is observed in germinal centers, in mantle zone cells, and in scattered cells of the inter-follicular areas. Anti-CD19 exhibits an overall immunoreactivity pattern similar to those of the antibodies against CD20 and CD22. However, in contrast to CD20, expression of CD19 is continuous throughout B-cell development and through terminal differentiation of B-cells into plasma cells. Anti-CD19 positivity is seen in the vast majority of B-cell neoplasms commonly at a lower intensity than normal B-cell counterparts. Plasma cell neoplasms are nearly always negative, as are T-cell neoplasms. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title