Learn More
CD19 Antibody (FMC63), Janelia Fluor™ 549, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP252716JF549
Catalog No. NBP52716549
Only null left
Add to Cart
Description
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence.
Specifications
Specifications
CD19
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD19
Human prolymphocytic leukaemia cell line JVM3.
0.1 mL
Primary
Binds human CD19.
Store at 4°C in the dark.
IgG2a κ
Flow Cytometry, Immunohistochemistry, Immunofluorescence
FMC63
Flow Cytometry, Immunohistochemistry, Immunofluorescence
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Mouse
Protein A or G purified
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers
930
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.