Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Antibody (HIB19), PerCP/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
$572.50
Specifications
Antigen | CD19 |
---|---|
Clone | HIB19 |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Specifications
CD19 | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Mouse | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
930 | |
The immunogen for this antibody was CD19. | |
Primary | |
Store at 4C. Do not freeze. |
HIB19 | |
Flow Cytometry | |
PerCP-Cyanine5.5 | |
RUO | |
Human | |
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
CD19 | |
IgG1 κ | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title