Learn More
CD19 Antibody (SJ25-C1), Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP128375
Catalog No. NBP128375
Only null left
Add to Cart
Description
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
CD19
Monoclonal
0.1 mg/mL
Flow Cytometry = 1 ug/10^6 cells, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
CD19
NALM-1 and NALM-16 leukemia cell line
0.1 mg
B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
930
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG1
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Blocking Assay
SJ25-C1
Unconjugated
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only