Learn More
CD19 Monoclonal Antibody (1G9), Exalpha

Mouse Monoclonal Antibody
Supplier: Vector Laboratories 0191100UG


Description
Flow Cytometry PBMC: Add 10 μL of MAB/10^6 PBMC in 100 μL PBS. Mix gently and incubate for 15 minutes at 2 to 8C. Wash twice with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. Flow Cytometry Whole Blood: Add10 μL of MAB/100 μL of whole blood. Mix gently and incubate for 15 minutes at room temperature 20C. Lyse the whole blood. Wash once with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. See instrument manufacturer instructions for Lysed Whole Blood and Immunofluorescence analysis with a flow cytometer or microscope.
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow CytometrySpecifications
CD19 | |
Monoclonal | |
Unconjugated | |
Cd19 | |
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
Protein A/G | |
RUO | |
930 | |
4° C, do not freeze | |
Liquid |
Flow Cytometry | |
1G9 | |
PBS with 0.075% sodium azide; pH 7.2 | |
P15391 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.