Learn More
Invitrogen™ CD19 Monoclonal Antibody (6D5)
Rat Monoclonal Antibody
$409.00 - $566.50
Specifications
Antigen | CD19 |
---|---|
Clone | 6D5 |
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA180359
|
Invitrogen™
MA180359 |
100 μg |
Each for $409.00
|
|
|||||
PIMA516534
|
Invitrogen™
MA516534 |
250 μg |
Each for $566.50
|
|
|||||
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse CD19 antibody, clone 6D5 recognizes the murine CD19 cell surface antigen, a 95kDa glycoprotein expressed by B lymphocytes. Clone 6D5 has been shown to recognize the same epitope as clone ID3 in cross-competition assays.
CD19 is a member of the immunoglobulin superfamily and has two Ig like domains. The CD19 molecule is expressed on 100% of the peripheral B cells as defined by expression of kappa or lambda light chains. CD19 appears to be expressed on myeloid leukemia cells, particularly those of monocytic lineage. Leukemia phenotype studies have demonstrated that the earliest and broadest B cell restricted antigen is the CD19 antigen. The receptor for CD19 is an important functional regulator of normal and malignant B cell proliferation, and is expressed in all B cell precursor leukemias. Lymphocytes proliferate and differentiate in response to various concentrations of different antigens. The ability of the B cell to respond in a specific, yet sensitive manner to the various antigens is achieved with the use of low-affinity antigen receptors. CD19 is a cell surface molecule which assembles with the antigen receptor of B lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation. Besides being a signal-amplifying coreceptor for the B cell receptor (BCR), CD19 can also signal independently of BCR co-ligation and is a central regulatory component upon which multiple signaling pathways converge. Mutation of the CD19 gene results in hypogammaglobulinemia, whereas CD19 overexpression causes B cell hyperactivity.Specifications
CD19 | |
1 mg/mL | |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
Unconjugated | |
Rat | |
RUO | |
PBS with 0.09% sodium azide; pH 7.4 | |
P25918 | |
12478 | |
Human K562 cell line transfected with murine CD19. | |
Antibody |
6D5 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG2a | |
Mouse | |
Cd19 | |
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
Cd19 | |
Primary | |
Protein G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.