Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Monoclonal Antibody (HIB19), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$209.00 - $463.00
Specifications
Antigen | CD19 |
---|---|
Clone | HIB19 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H004T03R03A
![]() |
Invitrogen™
H004T03R03A |
100 Tests |
Each for $463.00
|
|
|||||
H004T02R03A
![]() |
Invitrogen™
H004T02R03A |
25 Tests |
Each for $209.00
|
|
|||||
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow CytometrySpecifications
CD19 | |
5 μL/Test | |
Monoclonal | |
Mouse | |
Human | |
P15391 | |
930 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
HIB19 | |
Flow Cytometry | |
NovaFluor Red 700 | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
IgG1 κ | |
Affinity chromatography | |
Cd19 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title