Learn More
CD19 Mouse anti-Human, Biotin, Clone: COC19, Abnova™
Mouse monoclonal antibody raised against human CD19.
Supplier: Abnova Corporation MAB12401
Description
Lymphocytes proliferate and differentiate in response to various concentrations of different antigens. The ability of the B cell to respond in a specific, yet sensitive manner to the various antigens is achieved with the use of low-affinity antigen receptors. This gene encodes a cell surface molecule which assembles with the antigen receptor of B lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation. [provided by RefSeq]
Specifications
CD19 | |
Monoclonal | |
Biotin | |
Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user. | |
B4/MGC12802 | |
Mouse | |
Protein A/G affinity chromatography | |
RUO | |
930 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Flow Cytometry, Immunofluorescence | |
COC19 | |
Mouse monoclonal antibody raised against human CD19. | |
In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA) | |
CD19 | |
CD19 | |
50 Reactions | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.