Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Mouse anti-Human, BUV563, Clone: SJ25C1, BD Horizon™
Mouse Monoclonal Antibody
$248.00 - $472.00
Specifications
Antigen | CD19 |
---|---|
Clone | SJ25C1 (also known as SJ25-C1) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 563 |
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Specifications
CD19 | |
Flow Cytometry | |
Brilliant Ultraviolet 563 | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |
SJ25C1 (also known as SJ25-C1) | |
Monoclonal | |
Mouse | |
Human | |
Primary | |
CD19 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title