Learn More
CD19 Mouse anti-Human , BUV615, Clone: SJ25C1 (also known as SJ25-C1), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD19 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SJ25C1 (also known as SJ25-C1) |
| Conjugate | Brilliant Ultraviolet 615 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | B4; B-lymphocyte antigen CD19; Leu-12 |
| Host Species | Mouse |
| Immunogen | Human NALM1 + NALM16 cells |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.