Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Mouse anti-Human, Clone: 1C10A1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | CD19 |
|---|---|
| Clone | 1C10A1 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-106-0626
![]()
|
Proteintech Group Inc
662981IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
CD19 is a member of the immunoglobulin superfamily and has two Ig like domains. The CD19 molecule is expressed on 100% of the peripheral B cells as defined by expression of kappa or lamda light chains. It appears to be expressed on myeloid leukemia cells, particularly those of monocytic lineage. Leukemia phenotype studies have demonstrated that the earliest and broadest B cell restricted antigen is the CD19 antigen. The receptor for CD19 is an important functional regulator of normal and maligt B cell proliferation. It is expressed in all B cell precursor leukemias.Specifications
| CD19 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B lymphocyte antigen CD19, B4, CD19, CD19 molecule, Differentiation antigen CD19, T cell surface antigen Leu 12 | |
| CD19 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1C10A1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P15391 | |
| 930 | |
| CD19 Fusion Protein Ag19945 | |
| Primary | |
| -20°C | |
| CD19 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title