Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Mouse anti-Human, Clone: CAT-13.1E10, Novus Biologicals™

Mouse Monoclonal Antibody
$280.00 - $596.50
Specifications
Antigen | CD19 |
---|---|
Clone | CAT-13.1E10 |
Concentration | 1 mg/ml |
Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
Applications | Flow Cytometry, Immunofluorescence |
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
CD19 | |
1 mg/ml | |
Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
PBS | |
930 | |
IgG1 κ | |
Protein A purified |
CAT-13.1E10 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
Human | |
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
This antibody was raised by immunising mice against CD19. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title