Learn More
CD19 Mouse anti-Human, PE, Clone: HIB19, BD
Mouse Monoclonal Antibody
$74.00 - $158.70
Specifications
Antigen | CD19 |
---|---|
Clone | HIB19 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561741
|
BD Biosciences
561741 |
25 Tests |
Each for $74.00
|
|
|||||
BDB555413
|
BD Biosciences
555413 |
100 Tests |
Each for $158.70
|
|
|||||
Description
The HIB19 monoclonal antibody specifically binds to the 95 kDa type I transmembrane CD19 glycoprotein. CD19 is expressed during all stages of B-cell maturation and differentiation, except on plasma cells. CD19 is also present on follicular dendritic cells. It is not found on T cells or on normal granulocytes. CD19 is a signal transduction molecule that regulates B cell development, activation, proliferation and differentiation. It associates with the complement receptor 2 (CD21), TAPA-1 (CD81), Leu 13, and/or MHC class II to form a signal transduction complex on the surface of B cells. Anti-CD19 clone HIB19 partially blocks the binding of clone B43, another CD19-specific monoclonal antibody.
Specifications
CD19 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
HIB19 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.