Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Mouse anti-Human, R-PE, Clone: SJ25-C1, Southern Biotech™

Mouse Monoclonal Antibody
$83.80 - $201.50
Specifications
Antigen | CD19 |
---|---|
Clone | SJ25-C1 |
Concentration | Lot specific |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD19 | |
Lot specific | |
Monoclonal | |
Mouse | |
Human | |
NALM-1 and NALM-16 leukemia cell line | |
Primary |
SJ25-C1 | |
Flow Cytometry | |
R-PE | |
RUO | |
Supplied in PBS/NaN3 and a stabilizing agent | |
IgG1 | |
Store at 2-8°C; Protect from light; Do not freeze |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title