Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD193 (CCR3) Monoclonal Antibody (eBio5E8-G9-B4 (5E8-G9-B4)), eBioscience™, Invitrogen™

Catalog No. 14193937
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-193-937 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-193-937 Supplier Invitrogen™ Supplier No. 14193937
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The eBio5E8-G9-B4 MAb reacts with human CD193 (CCR3, CKR3). CD193 is a member of the seven transmembrane G-protein coupled receptor (GPCR) family, and is a high affinity chemokine receptor for the chemokines eotaxin-1 (CCL11), eotaxin-2 (CCL24), eotaxin-3 (CCL26) and MCP-4 (CCL13), but has also been reported to bind RANTES, MCP-3 and MCP-4. CD193 is highly expressed on the surface of eosinophils, and is the key eosinophil chemokine receptor responsible for the regulation of eosinophil migration and function. In addition to eosinophils, CD193 is also expressed on CD4+ TH2 cells, basophils, mast cells, mononuclear phagocytes, platelets, CD34+ hematopoietic progenitors and airway epithelial cells. CD193 likely plays an important role in the migration of eosinophils in allergic airway inflammation and asthma. CD193 can also serve as a receptor for HIV-1 cell entry.

CCR3 (C-C chemokine receptor type 3) binds to eotaxin, eotaxin-3, MCP-3, MCP-4, RANTES, and MIP-1 delta. CCR3 trasnduces a signal by increasing the intracellular calcium ions level. It also serves as an alternative co-receptor with CD4 for HIV-1 infection. Other chemokines play important roles in inflammation and are critical for the recruitment of effector immune cells to sites of infection. Chemokines activate leukocytes by binding to G protein coupled receptors. The ever-growing chemokine receptor subtypes can be divided into 2 major groups, CXCR and CCR, based on the 2 major classes of chemokines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD193 (CCR3)
Applications Flow Cytometry
Classification Monoclonal
Clone eBio5E8-G9-B4 (5E8-G9-B4)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CCR3
Gene Accession No. P51677
Gene Alias b-chemokine receptor; CC chemokine receptor 3; C-C chemokine receptor type 3; C-C CKR-3; C-C motif chemokine receptor 3; CC-CKR3; CC-CKR-3; Ccr3; CCR-3; CD193; chemokine (C-C motif) receptor 3; chemokine (C-C) receptor 1,-like 2; chemokine (C-C) receptor 3; CKR3; Cmkbr1l2; CMKBR3; eosinophil CC chemokine receptor 3; eosinophil eotaxin receptor; Macrophage inflammatory protein 1-alpha receptor-like 2; MGC102841; MIP-1 alpha RL2; MIP-1 alphaRL2; probable C-C chemokine receptor type 3
Gene Symbols CCR3
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1232
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.