Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD193 (CCR3) Rat anti-Mouse, BV510, Clone: 83103, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 747820
Description
CD193 (CCR3) Monoclonal antibody specifically detects CD193 (CCR3) in Mouse samples. It is validated for Flow Cytometry.
Specifications
CD193 (CCR3) | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
83103 | |
Brilliant Violet 510 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction