Learn More
CD195 (CCR5) Monoclonal Antibody (HM-CCR5 (7A4)), PerCP-eFluor™ 710, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody
$174.00 - $427.00
Specifications
Antigen | CD195 (CCR5) |
---|---|
Clone | HM-CCR5 (7A4) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-3789
![]() |
Invitrogen™
46195180 |
25 μg |
Each for $174.00
|
|
|||||
50-112-3790
![]() ![]() |
Invitrogen™
46195182 |
100 μg |
Each for $427.00
|
![]() |
|||||
Description
Description: CCR5 is the major co-receptor for macrophage-tropic isolates of HIV-1. CCR5, also known as CD195, is a seven transmembrane chemokine receptor capable of binding RANTES, MIP-1 alpha and MIP-1 beta. Knockout mice show a decreased recruitment of macrophages and increased susceptibility to infection. Protein and mRNA has been found in macrophages from thioglycolate-treated and activated T cells. The HM-CCR5(7A4) monoclonal antibody binds the N terminal extracellular domain of mouse CCR5 with no crossreactivity to human CCR5. Applications Reported: This HM-CCR5 (7A4) antibody has been reported for use in flow cytometric analysis. Applications Tested: This HM-CCR5 (7A4) antibody has been tested by flow cytometric analysis of mouse thioglycolated- induced peritoneal exudate cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome.
CD195 (CCR5) Monoclonal antibody specifically detects CD195 (CCR5) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD195 (CCR5) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AM4-7; C-C chemokine receptor type 5; C-C CKR-5; C-C motif chemokine receptor 5 (gene/pseudogene); C-C motif chemokine receptor 5 A159A; CCCKR5; CC-CKR-5; Ccr5; CCR-5; CD195; CD195 antigen; chemokine (C-C motif) receptor 5; chemokine (C-C) receptor 5; chemokine receptor CCR5; chemr13; CKR5; CKR-5; Cmkbr5; HIV-1 fusion coreceptor; IDDM22; MIP-1 alpha receptor | |
CCR5 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
CCR5 |
HM-CCR5 (7A4) | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Armenian Hamster | |
Mouse | |
P51682 | |
12774 | |
IgG | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.