Learn More
Invitrogen™ CD195 (CCR5) Monoclonal Antibody (NP-6G4), APC, eBioscience™

Mouse Monoclonal Antibody
$375.00
Specifications
Antigen | CD195 (CCR5) |
---|---|
Clone | NP-6G4 |
Concentration | 5 μL/Test |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
17-195-642
![]() ![]() |
Invitrogen™
17195642 |
100 Tests |
Each of 1 for $375.00
|
![]() |
|||||
Description
Description: The NP-6G4 monoclonal antibody reacts with human CD195 (CCR5), an approximately 45 kDa member of the G protein-coupled receptor (GPCR) superfamily that binds chemokines including MIP-1a, MIP-1b and RANTES. Human CCR5 is expressed on resting T cells with a memory/effector phenotype, monocytes, macrophages and immature dendritic cells. CCR5 functions as a receptor to direct the chemotaxis of lymphocytes, and is the primary co-receptor for macrophage-tropic Human Immunodeficiency Virus Type 1 (HIV-1), which binds CCR5 through gp120. Applications Reported: This NP-6G4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This NP-6G4 antibody has been pre-titrated and tested on normal human peripheral blood cells. For optimal performance, it is recommended that staining with NP-6G4 be done at 37°C for 30 to 60 minutes in Flow Cytometry Staining Buffer. This antibody can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Human immunodeficiency virus (HIV) and related virus require coreceptors, in addition to CD4, to infect target cells. Some G protein-coupled receptors including CCR5, CXCR4, CCR3, CCR2b and CCR8 in the chemokine receptor family, and four new human molecules GPR15, STRL33, GPR1 and V28 were recently identified as HIV coreceptors. Among them, CCR5 (CC-CKR-5) is a principal coreceptor for macrophage- and dual-tropic HIV-1 strains fusion and entry of human white blood cells. CCR5 is required for the infection by HIV-1, HIV-2, and SIV. The beta-chemokines RANTES, MIP-alpha and MIP-beta are the ligands for CCR5 and prevent infection by M-tropic HIV-1. CCR5 associates with the surface CD4-gp120 of HIV complex and leads to membrane fusion and virus entry of target cells. The amino-terminal domain and the extracellular loops of CCR5 serve as HIV biding sites. CCR5 messenger RNA is expressed in lymphoid organs and monocytes.Specifications
CD195 (CCR5) | |
5 μL/Test | |
Flow Cytometry | |
APC | |
Mouse | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P51681 | |
1234 | |
Primary | |
Affinity chromatography |
NP-6G4 | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
CCR5 | |
AM4-7; C-C chemokine receptor type 5; C-C CKR-5; C-C motif chemokine receptor 5 (gene/pseudogene); C-C motif chemokine receptor 5 A159A; CCCKR5; CC-CKR-5; Ccr5; CCR-5; CD195; CD195 antigen; chemokine (C-C motif) receptor 5; chemokine (C-C) receptor 5; chemokine receptor CCR5; chemr13; CKR5; CKR-5; Cmkbr5; HIV-1 fusion coreceptor; IDDM22; MIP-1 alpha receptor | |
CCR5 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.