Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD195 (CCR5) Monoclonal Antibody (NP-6G4), PE, eBioscience™
GREENER_CHOICE

Catalog No. 501122293
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-2293 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-2293 Supplier Invitrogen™ Supplier No. 12195642
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: The NP-6G4 monoclonal antibody reacts with human CD195 (CCR5), an approximately 45 kDa member of the G protein-coupled receptor (GPCR) superfamily that binds chemokines including MIP-1a, MIP-1b and RANTES. Human CCR5 is expressed on resting T cells with a memory/effector phenotype, monocytes, macrophages and immature dendritic cells. CCR5 functions as a receptor to direct the chemotaxis of lymphocytes, and is the primary co-receptor for macrophage-tropic Human Immunodeficiency Virus Type 1 (HIV-1), which binds CCR5 through gp120. Applications Reported: This NP-6G4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This NP-6G4 antibody has been pre-titrated and tested on normal human peripheral blood cells. For optimal performance, it is recommended that staining with NP-6G4 be done at 37°C for 1 hour in Flow Cytometry Staining Buffer (Product # 00-4222-μ). This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: μ8 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Human immunodeficiency virus (HIV) and related virus require coreceptors, in addition to CD4, to infect target cells. Some G protein-coupled receptors including CCR5, CXCR4, CCR3, CCR2b and CCR8 in the chemokine receptor family, and four new human molecules GPR15, STRL33, GPR1 and V28 were recently identified as HIV coreceptors. Among them, CCR5 (CC-CKR-5) is a principal coreceptor for macrophage- and dual-tropic HIV-1 strains fusion and entry of human white blood cells. CCR5 is required for the infection by HIV-1, HIV-2, and SIV. The beta-chemokines RANTES, MIP-alpha and MIP-beta are the ligands for CCR5 and prevent infection by M-tropic HIV-1. CCR5 associates with the surface CD4-gp120 of HIV complex and leads to membrane fusion and virus entry of target cells. The amino-terminal domain and the extracellular loops of CCR5 serve as HIV biding sites. CCR5 messenger RNA is expressed in lymphoid organs and monocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD195 (CCR5)
Applications Flow Cytometry
Classification Monoclonal
Clone NP-6G4
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CCR5
Gene Accession No. P51681
Gene Alias AM4-7; C-C chemokine receptor type 5; C-C CKR-5; C-C motif chemokine receptor 5 (gene/pseudogene); C-C motif chemokine receptor 5 A159A; CCCKR5; CC-CKR-5; Ccr5; CCR-5; CD195; CD195 antigen; chemokine (C-C motif) receptor 5; chemokine (C-C) receptor 5; chemokine receptor CCR5; chemr13; CKR5; CKR-5; Cmkbr5; HIV-1 fusion coreceptor; IDDM22; MIP-1 alpha receptor
Gene Symbols CCR5
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1234
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.