Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD195 Mouse anti-Human,Cynomolgus,Rhesus Ma, BB515, Clone: 3A9, BD Horizon™

Catalog No. BDB564512 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564512 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564512 Supplier BD Biosciences Supplier No. 564512
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD195 Monoclonal antibody specifically detects CD195 in Human,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD195
Applications Flow Cytometry
Classification Monoclonal
Clone 3A9
Conjugate Brilliant Blue 515 (BB515)
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Cynomolgus, Rhesus Macaque
Content And Storage Refrigerate (2 to 8°C)
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.