Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD195 Mouse anti-Human, PE-Cy5, Clone: 2D7/CCR5, BD

Catalog No. BDB561750 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561750 25 Tests
BDB556889 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561750 Supplier BD Biosciences Supplier No. 561750

Mouse Monoclonal Antibody

The 2D7/CCR5 monoclonal antibody specifically binds to the human chemokine receptor, CCR5. CCR5 is also known as CD195. CCR5 is a seven transmembrane-spanning G protein-associated molecule. CCR5 belongs to the beta chemokine receptor family. It is expressed on a subset of T lymphocytes (CD3+CD45RO+CD95+). CCR5 regulates lymphocyte chemotaxis activation and transendothelial migration during inflammation. It signals a response to at least three chemokines: Regulated upon Activation Normal T-cell Expressed and Secreted (RANTES), Macrophage Inflammatory Protein-1 (MIP-1) and Monocyte Chemoattractant Protein 2 (MCP-2). Additionally, CCR5 has been found to be a coreceptor for macrophage-tropic HIV-1 on CD4+ cells, a characteristic that is important in viral transmission. Reports indicate that individuals who have partial (heterozygous) or complete (homozygous) deletion of the CCR5 allele, demonstrate resistance to HIV infection. This antibody has been shown to block ligand and gp120 binding. It is also able to neutralize HIV infection.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD195
Applications Flow Cytometry
Classification Monoclonal
Clone 2D7/CCR5
Conjugate PE-Cyanine5
Description CCR5; C-C chemokine receptor type 5; CC-CKR-5; CKR5; CHEMR13
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CCR5 Transfected Cell Line
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.