Learn More
CD196 (CCR6) Mouse anti-Human, Baboon, Cynomolgus, Rhesus, BB700, Clone: 11A9, BD Horizon™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD196 (CCR6) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 11A9 |
| Conjugate | Brilliant Blue 700 (BB700) |
| Host Species | Mouse |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Test Specificity | CD196 (CCR6) |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.