Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD197, CCR7 Rat anti-Human, APC-R700, Clone: 3D12, BD Horizon™

Catalog No. BDB565867 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565867 100 Tests
BDB565868 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB565867 Supplier BD Biosciences Supplier No. 565867
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD197, CCR7 Monoclonal antibody specifically detects CD197, CCR7 in Human samples. It is validated for Flow cytometry.

CD197, CCR7 Monoclonal antibody specifically detects CD197, CCR7 in Human samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD197, CCR7
Applications Flow Cytometry
Classification Monoclonal
Clone 3D12
Conjugate APC-R700
Description Vol. Per test: 5μL
Formulation Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide.
Gene Alias CCR7, BLR-2, EBI-1, CMKBR7
Host Species Rat
Immunogen Human CCR7 protein
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.