Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD1a Ab-2, Mouse Monoclonal Antibody

Catalog No. MS746S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS746S 1 mL Concentrated
MS746S1 500 μL Concentrated
MS746S0 100 μL Concentrated
MS746R1 1 mL Prediluted
MS746R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS746S Supplier Epredia™ Supplier No. MS746S

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD1a Ab-2, Mouse Monoclonal Antibody.

At least five CD1 genes (CD1a, b, c, d, and e) have been identified. CD1 is expressed on cortical thymocytes, Langerhans cells, and dendritic cells. It is absent on mature peripheral blood T cells but intracytoplasmic expression is detected on activated T lymphocytes. CD1 proteins have been demonstrated to restrict T-cell response to non-peptide lipid and glycolipid antigens and play a role in non-classical antigen presentation.

Host Species: Mouse

Clone: MTB1

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: External domain

Immunogen: Full external domain of CD1a

Molecular Weight: 49 kDa

Positive Control: Thymus

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD1a Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MTB1
Conjugate Unconjugated
Host Species Mouse
Immunogen Full external domain of CD1a
Purification Method Thymus
Quantity 1 mL
Regulatory Status RUO
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.