Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1a Antibody (201B5.08), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0081A647100
Description
CD1a Monoclonal specifically detects CD1a in Human, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence.Specifications
CD1a | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunofluorescence | |
CD1A | |
CD1a was used as immunogen. | |
0.1 mg | |
Dendritic Cell Markers, Immunology | |
909 | |
Store at -20C. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
201B5.08 | |
Alexa Fluor 647 | |
CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
Human, Pig | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction