Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1a Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$382.00 - $610.00
Specifications
Antigen | CD1a |
---|---|
Concentration | 0.1mg/mL |
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CD1a Polyclonal specifically detects CD1a in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CD1a | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Unconjugated | |
RUO | |
Human | |
CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
CD1A | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
0.1mg/mL | |
Polyclonal | |
Rabbit | |
Dendritic Cell Markers, Immunology | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
909 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:QNLVSGWLSDLQTHTWDSNSSTIVFLCPWSRGNFSNEEWKELETLFRIRTIRSFEGIRRYAHELQFEYPFEIQVTGGCELHSGKVSGSFLQLAYQGSDFVSFQNNSWLPYPVAGNMAKHFCKVLNQNQHEND | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title