Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1a Mouse, anti-Human, R718, Clone: SK9, BD OptiBuild™

Catalog No. BDB751847 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751847 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751847 Supplier BD Biosciences Supplier No. 751847
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The SK9 monoclonal antibody specifically recognizes CD1a. CD1a is a ∼49 kDa type I transmembrane glycoprotein that associates non-covalently with β2-microglobulin. The CD1a antigen is present on 60% to 90% of thymocytes, some T-cell leukemias and lymphomas, Langerhans cells, and a subset of dendritic cells. The CD1a antibody does not react with peripheral blood T and B lymphocytes, monocytes, normal bone marrow mononuclear cells, or normal tonsillar B and T lymphocytes. The CD1a antigen has structural similarities to MHC class I antigens and plays a role in the presentation of non-peptide antigens.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD1a
Applications Flow Cytometry
Classification Monoclonal
Clone SK9
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.