Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1a Rabbit anti-Human, Clone: CBT6, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | CD1a |
---|---|
Clone | CBT6 |
Dilution | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CD1a Monoclonal antibody specifically detects CD1a in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
CD1a | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
RUO | |
Human | |
CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
This antibody was raised by immunizing mice with the human CD1a protein. | |
Primary |
CBT6 | |
Monoclonal | |
Rabbit | |
Dendritic Cell Markers, Immunology | |
PBS | |
909 | |
IgG κ | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title