Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1b Antibody (CC20), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065315IR
Description
CD1b Monoclonal specifically detects CD1b in Bovine, Canine, Equine, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD1b | |
Monoclonal | |
DyLight 755 | |
CD1, CD1A, CD1b antigen, CD1B antigen, b polypeptide, CD1b molecule, cortical thymocyte antigen CD1B, differentiation antigen CD1-alpha-3, MGC125990, MGC125991, R1, T-cell surface glycoprotein CD1b | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Recognizes the bovine CD1w2 cell surface antigen, a glycoprotein heterodimer of 12kD and 46kD. CD1w2 is expressed by cortical thymocytes and a minority of medullary thymocytes, with a pattern similar to antibodies of the CD1b cluster in humans. | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
CC20 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
CD1B | |
Made to Bovine CD1w2 | |
0.125 mL | |
Immunology | |
910 | |
Bovine, Canine, Equine, Goat, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction