Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1b/c Antibody (B-B5), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318096AF750
Description
CD1b/c Monoclonal antibody specifically detects CD1b/c in Human samples. It is validated for Flow CytometrySpecifications
CD1b/c | |
Monoclonal | |
Alexa Fluor 750 | |
BDCA1, CD1, CD1A, CD1B antigen, b polypeptide, CD1C antigen, c polypeptide, cortical thymocyte antigen CD1B, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R1, R7, T-cell surface glycoprotein CD1b, T-cell surface glycoprotein CD1c | |
Thymus cells and Jurkat cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells | |
0.1 mL | |
CD Markers | |
910 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
B-B5 | |
50mM Sodium Borate | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction