Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1b Mouse anti-Human, Guinea Pig, MilliporeSigma™

Catalog No. MABF203125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2031-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2031-25 Supplier MilliporeSigma Supplier No. MABF203125UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD1b, clone BCD1b3.1 is a mouse monoclonal antibody that detects T-cell surface glycoprotein CD1b and it has been tested for use in Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition assays, and Neutralizing function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD1b
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition Assays, Neutralization
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CD1B
Host Species Mouse
Immunogen Isolated human CD1+ monocytes.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone BCD1b3.1 detects T-cell surface glycoprotein CD1b in human and guinea pig.
Target Species Human, Guinea Pig
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.