Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD1b3 Monoclonal Antibody (MsGP9), PE
GREENER_CHOICE

Catalog No. PIMA516873
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516873 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516873 Supplier Invitrogen™ Supplier No. MA516873
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD1b3
Applications Flow Cytometry
Classification Monoclonal
Clone MsGP9
Concentration Conc. Not Determined
Conjugate PE
Formulation PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
Gene Cd1b3
Gene Accession No. Q9QZZ0
Gene Alias Cd1b3; CD1-b3; CD1b-3; CD1b3 molecule; GP-CD1-B3; T-cell surface glycoprotein CD1b3
Gene Symbols Cd1b3
Host Species Mouse
Immunogen Guinea pig thymocytes.
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100379551
Target Species Guinea Pig
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.