Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1c/BDCA-1 Antibody (L161), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262220AF750
Description
CD1c/BDCA-1 Monoclonal specifically detects CD1c/BDCA-1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
CD1c/BDCA-1 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD1C | |
human thymocytes | |
0.1 mL | |
Primary | |
The antibody MEM-97 reacts with CD20 (Bp35), a 33-37 kDa non-glycosylated membrane receptor with four transmembrane domains, expressed on B lymphocytes (it is lost on plasma cells), follicular dendritic cells, and at low levels on peripheral blood T lymphocytes. | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry | |
L161 | |
Flow Cytometry, Immunohistochemistry | |
BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c | |
Mouse | |
Protein A purified | |
Cancer, Immunology | |
911 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction