Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1c/BDCA-1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298701MFV610
Description
CD1c/BDCA-1 Polyclonal antibody specifically detects CD1c/BDCA-1 in Human samples. It is validated for Western BlotSpecifications
CD1c/BDCA-1 | |
Polyclonal | |
Western Blot | |
BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c | |
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human CD1c/BDCA-1. | |
0.1 mL | |
Cancer, Immunology | |
911 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction