Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD1c Monoclonal Antibody (3G1B3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548408
Description
CD1c Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA
This gene encodes a member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by this gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail, and requires vesicular acidification to bind lipid antigens.
Specifications
CD1c | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P29017 | |
CD1C | |
Purified recombinant fragment of human CD1C (AA: extra 18-302) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
3G1B3 | |
Unconjugated | |
CD1C | |
BDCA1; canCD1c; CD1; CD1A; CD1C; CD1C antigen, c polypeptide; CD1c molecule; cortical thymocyte antigen CD1C; differentiation antigen CD1-alpha-3; R7; RP11-101J8.3; T-cell surface glycoprotein CD1c | |
Mouse | |
Protein G | |
RUO | |
911 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction