Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1d Antibody (1B1), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143461AF350
Description
CD1d Monoclonal specifically detects CD1d in Mouse, Hamster samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
CD1d | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CD1D | |
The immunogen for this antibody was CD1d. | |
Primary | |
Mouse, Hamster | |
IgG2b κ |
Flow Cytometry, Immunohistochemistry | |
1B1 | |
Flow Cytometry, Immunohistochemistry | |
antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D | |
Rat | |
0.1 mL | |
912 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction