Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1d Antibody (51.1), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143460JF646
Description
CD1d Monoclonal specifically detects CD1d in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD1d | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD1D | |
The immunogen for this antibody was CD1d. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
51.1 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D | |
Mouse | |
Protein A or G purified | |
Alzheimers Research, B Cell Development and Differentiation Markers, Cellular Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Neuroscience | |
912 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction