Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1d Mouse anti-Mouse, BUV737, Clone: WTH2 (also known as WTH-2), BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 749068
Description
CD1d Monoclonal antibody specifically detects CD1d in Mouse samples. It is validated for Flow Cytometry
Specifications
CD1d | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
Cd1d; Antigen-presenting glycoprotein CD1d; CD1d1 | |
Rat CD1d Transfected Cell Line | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
WTH2 (also known as WTH-2) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction