Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1d Mouse, anti-Rat, R718, Clone: WTH2, BD OptiBuild™

Catalog No. BDB751970 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751970 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751970 Supplier BD Biosciences Supplier No. 751970
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The WTH2 monoclonal antibody specifically binds to rat CD1d and crossreacts with mouse CD1d. CD1d is a type I transmembrane glycoprotein that is noncovalently associated with β2-microglobulin. CD1d is expressed on various cell types including subsets of thymocytes, T cells, B cells, monocytes, macrophages, dendritic cells, endothelial cells and epithelial cells. Although structurally similar to MHC Class I antigens, CD1d molecules are rather non-polymorphic and serve to present non-peptide antigens such as endogenous or microbial glycolipids to T lymphocytes (NKT cells). The WTH1 monoclonal antibody reportedly binds to a non-overlapping epitope on rat CD1d and also crossreacts with mouse CD1d when compared with WTH2. Both WTH1 and WTH2 reportedly interfere with antigen recognition by CD1d-restricted T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD1d
Applications Flow Cytometry
Classification Monoclonal
Clone WTH2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.