Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1e Antibody (704407), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7330H
Description
CD1e Monoclonal antibody specifically detects CD1e in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| CD1e | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Immunology | |
| 913 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 704407 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CD1e molecule, e polypeptide, FLJ17609, membrane-associated, thymocyte antigen CD1E | |
| Mouse myeloma cell line NS0-derived recombinant human CD1e, Met1-Tyr304, Accession # P15812 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction