Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1e Mouse anti-Human, PE, Clone: 20.6 (also known as 20.6), BD Pharmingen™
Mouse Monoclonal Antibody
$218.00 - $430.00
Specifications
Antigen | CD1e |
---|---|
Clone | 20.6 (also known as 20.6) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD1e Monoclonal antibody specifically detects CD1e in Human samples. It is validated for Intracellular staining (Flow Cytometry)Specifications
CD1e | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human CD1e Transfected Cell Line | |
Primary |
20.6 (also known as 20.6) | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
CD1E; hCD1e; R2 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title