Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1E Polyclonal Antibody, FITC, Bioss
SDP

Catalog No. 502281066
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1066 Supplier Bioss Supplier No. BS23497RFITC
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD1E Polyclonal antibody specifically detects CD1E in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CD1E
Applications Flow Cytometry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene CD1E
Gene Accession No. P15812
Gene Alias CD1A; CD1e; CD1E antigen, e polypeptide; CD1e molecule; differentiation antigen CD1-alpha-3; hCD1e; leukocyte differentiation antigen; R2; R2G1; sCD1e; T-cell surface glycoprotein CD1e, membrane-associated; T-cell surface glycoprotein CD1e, soluble; thymocyte antigen CD1E
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CD1E
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 913
Target Species Human
Content And Storage -20° C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.