Learn More
CD2 Antibody (12-15), PE, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP126669
Catalog No. NBP126669
Only null left
Add to Cart
Description
Description
CD2 Monoclonal specifically detects CD2 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, In vitro assay.
Specifications
Specifications
CD2
Monoclonal
0.1 mg/mL
Flow Cytometry <= 0.5 ug/10^6 cells, Immunoprecipitation 1:10-1:500, In vitro assay
CD2
Crude plasma membranes of ESb cells
0.2 mg
Primary
Mouse CD2/LFA-2 (Mr 45-58 kDa)
Store at 4C. Do not freeze.
IgG1 κ
Flow Cytometry, Immunoprecipitation, In vitro Assay
12-15
PE
CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5
Rat
Protein A or G purified
RUO
914
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only