Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD2 Antibody (299808), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB18562K
This item is not returnable.
View return policy
Description
CD2 Monoclonal antibody specifically detects CD2 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| CD2 | |
| Monoclonal | |
| DyLight 488 | |
| CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
| Mouse myeloma cell line NS0-derived recombinant human CD2, Lys25-Asp209, Accession # P06729 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Immunology, Myeloid derived Suppressor Cell, Stem Cells | |
| 914 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| 299808 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction