Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD2 Antibody (TS2/18), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222537G
Description
CD2 Monoclonal specifically detects CD2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
CD2 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD2 | |
Human LFA-2 (CD2) | |
0.2 mL | |
Adaptive Immunity, Apoptosis, Immunology | |
914 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry | |
TS2/18 | |
Western Blot, Flow Cytometry | |
CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction