Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD2 Antibody (YTH 655), Alexa Fluor™ 647, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP281042AF647
Description
CD2 Monoclonal specifically detects CD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CD2 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
CD2 | |
Recombinant monoclonal antibody to CD2. | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Immunology | |
914 | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
YTH 655 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
Rat | |
Protein A purified | |
RUO | |
Primary | |
This antibody specifically recognizes human CD2, a cell adhesion molecule found on the surface of T cells and natural killer (NK) cells. | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction