Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD2 Antibody (YTH 655), mFluor Violet 500 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP281042MFV500
Description
CD2 Monoclonal antibody specifically detects CD2 in Human samples. It is validated for Flow CytometrySpecifications
CD2 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rat | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
YTH 655 | |
Flow Cytometry | |
CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
CD2 | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Immunology, Myeloid derived Suppressor Cell, Stem Cells | |
914 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction