Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD2 or CD6 Predicted Antibody (LH43A), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261101
Description
CD2 or CD6 Predicted Monoclonal specifically detects CD2 or CD6 Predicted in Llama, Alpaca samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD2 or CD6 Predicted | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
Mouse | |
Unpurified | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Functional Assay | |
LH43A | |
Unconjugated | |
Ascites with 0.09% Sodium Azide | |
Llama mononuclear cells | |
0.1 mg | |
Llama, Alpaca | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction